ADVH49B8261B AChemBlock

tert-butyl N-(4-chloro-3-methoxy-2-pyridyl)carbamate

  • Molecular Weight : 258.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-4-chloro-3-methoxy-2-pyridyl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84535
Availability details for
Quantity
  • Availability
  • Add to Cart