ADVH49B8261C AChemBlock

tert-butyl N-(4,4-difluoropiperidin-3-yl)-N-methylcarbamate

  • Molecular Weight : 250.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-4-4-difluoropiperidin-3-yl-n-methylcarbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84536
Availability details for
Quantity
  • Availability
  • Add to Cart