ADVH49B8261D AChemBlock

ethyl 2-(piperidin-4-yl)acetate hydrochloride

  • Cas Number : 169458-04-2
  • Molecular Weight : 207.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-2-piperidin-4-yl-acetate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78708
Availability details for
Quantity
  • Availability
  • Add to Cart