ADVH49B8261E AChemBlock

3-(6-chloro-2-methoxypyridin-3-yl)-3-methylpiperidin-2-one

  • Cas Number : 1693763-28-8
  • Molecular Weight : 254.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-6-chloro-2-methoxypyridin-3-yl-3-methylpiperidin-2-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86623
Availability details for
Quantity
  • Availability
  • Add to Cart