ADVH49B8261F AChemBlock

N-(3-Methoxyphenyl)-4-nitrobenzenesulfonaMid

  • Cas Number : 16936-99-5
  • Molecular Weight : 308.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-3-methoxyphenyl-4-nitrobenzenesulfonamid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86784
Availability details for
Quantity
  • Availability
  • Add to Cart