ADVH49B82620 AChemBlock

2,2-difluoro-1-(6-methoxy-2-pyridyl)ethanol

  • Cas Number : 1692203-14-7
  • Molecular Weight : 189.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-2-difluoro-1-6-methoxy-2-pyridyl-ethanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78713
Availability details for
Quantity
  • Availability
  • Add to Cart