ADVH49B82635 AChemBlock

(1S,2S)-2-methoxycyclohexanamine;hydrochloride

  • Cas Number : 1668594-28-2
  • Molecular Weight : 165.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1s-2s-2-methoxycyclohexanamine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78755
Availability details for
Quantity
  • Availability
  • Add to Cart