ADVH49B82636 AChemBlock

tert-butyl cis-3-ethyl-4-hydroxypyrrolidine-1-carboxylate

  • Molecular Weight : 215.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-cis-3-ethyl-4-hydroxypyrrolidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84562
Availability details for
Quantity
  • Availability
  • Add to Cart