ADVH49B8263D AChemBlock

tert-butyl cis-2-oxo-octahydro-1H-pyrrolo[3,2-c]pyridine-5-carboxylate

  • Molecular Weight : 240.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-cis-2-oxo-octahydro-1h-pyrrolo-3-2-c-pyridine-5-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84570
Availability details for
Quantity
  • Availability
  • Add to Cart