ADVH49B8263F AChemBlock

3-bromo-4-methyl-4H-1,2,4-triazole

  • Cas Number : 16681-73-5
  • Molecular Weight : 161.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-bromo-4-methyl-4h-1-2-4-triazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78756
Availability details for
Quantity
  • Availability
  • Add to Cart