ADVH49B82644 AChemBlock

tert-butyl 7-hydroxy-4-azaspiro[2.5]octane-4-carboxylate

  • Molecular Weight : 227.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-7-hydroxy-4-azaspiro-2-5-octane-4-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84577
Availability details for
Quantity
  • Availability
  • Add to Cart