ADVH49B82645 AChemBlock

4,6-dichloropyridine-3-carbonitrile

  • Cas Number : 166526-03-0
  • Molecular Weight : 173

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-6-dichloropyridine-3-carbonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71636
Availability details for
Quantity
  • Availability
  • Add to Cart