ADVH49B82648 AChemBlock

3-chloro-2-methyl-6-(trifluoromethyl)quinoxaline

  • Cas Number : 166402-14-8
  • Molecular Weight : 246.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-chloro-2-methyl-6-trifluoromethyl-quinoxaline.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78763
Availability details for
Quantity
  • Availability
  • Add to Cart