ADVH49B8264B AChemBlock

(3R)-8-Fluoro-1-methyl-3-[2-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-2,4(1H,3H)-quinazolinedione

  • Cas Number : 1643446-66-5
  • Molecular Weight : 410.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3r-8-fluoro-1-methyl-3-2-methyl-3-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-phenyl-2-4-1h-3h-quinazolinedione.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78822
Availability details for
Quantity
  • Availability
  • Add to Cart