ADVH49B8264D AChemBlock

(2R)-5-bromo-6-fluoro-2-(2-hydroxypropan-2-yl)-2,3,4,9-tetrahydro-1H-carbazole-8-carboxamide

  • Cas Number : 1643156-22-2
  • Molecular Weight : 369.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2r-5-bromo-6-fluoro-2-2-hydroxypropan-2-yl-2-3-4-9-tetrahydro-1h-carbazole-8-carboxamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78825
Availability details for
Quantity
  • Availability
  • Add to Cart