ADVH49B8264E AChemBlock

tert-butyl 7-amino-2-oxa-5-azabicyclo[2.2.2]octane-5-carboxylate

  • Molecular Weight : 228.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-7-amino-2-oxa-5-azabicyclo-2-2-2-octane-5-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84587
Availability details for
Quantity
  • Availability
  • Add to Cart