ADVH49B82651 AChemBlock

4-bromo-7-ethoxycarbonyl-3-fluoro-6,7,8,9-tetrahydro-5H-carbazole-1-carboxylic acid

  • Cas Number : 1643156-19-7
  • Molecular Weight : 384.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-bromo-7-ethoxycarbonyl-3-fluoro-6-7-8-9-tetrahydro-5h-carbazole-1-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78827
Availability details for
Quantity
  • Availability
  • Add to Cart