ADVH49B82654 AChemBlock

1-tert-butyl 2-methyl 1,2,3,6-tetrahydropyridine-1,2-dicarboxylate

  • Cas Number : 164298-41-3
  • Molecular Weight : 241.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-tert-butyl-2-methyl-1-2-3-6-tetrahydropyridine-1-2-dicarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78830
Availability details for
Quantity
  • Availability
  • Add to Cart