ADVH49B82657 AChemBlock

3-bromo-7H-pyrrolo[2,3-c]pyridazine

  • Cas Number : 1638764-04-1
  • Molecular Weight : 198.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-bromo-7h-pyrrolo-2-3-c-pyridazine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78907
Availability details for
Quantity
  • Availability
  • Add to Cart