ADVH49B82659 AChemBlock

4-Fluoro-1H-indazole-3-carbonitrile

  • Cas Number : 1638591-51-1
  • Molecular Weight : 161.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-fluoro-1h-indazole-3-carbonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72418
Availability details for
Quantity
  • Availability
  • Add to Cart