ADVH49B827B1 AChemBlock

tert-butyl 4-(hydroxymethyl)-2-azabicyclo[2.2.2]octane-2-carboxylate

  • Molecular Weight : 241.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4-hydroxymethyl-2-azabicyclo-2-2-2-octane-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84629
Availability details for
Quantity
  • Availability
  • Add to Cart