ADVH49B827B4 AChemBlock

tert-butyl 3-methyl-5-oxo-2-azabicyclo[4.1.0]heptane-2-carboxylate

  • Molecular Weight : 225.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-methyl-5-oxo-2-azabicyclo-4-1-0-heptane-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84634
Availability details for
Quantity
  • Availability
  • Add to Cart