ADVH49B827B9 AChemBlock

tert-butyl 3-amino-3-(2-ethoxy-1-fluoro-2-oxo-ethyl)azetidine-1-carboxylate

  • Molecular Weight : 276.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-amino-3-2-ethoxy-1-fluoro-2-oxo-ethyl-azetidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84646
Availability details for
Quantity
  • Availability
  • Add to Cart