ADVH49B827BB AChemBlock

tert-butyl 3-amino-3-(1-fluoro-2-hydroxy-ethyl)azetidine-1-carboxylate

  • Molecular Weight : 234.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-amino-3-1-fluoro-2-hydroxy-ethyl-azetidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84648
Availability details for
Quantity
  • Availability
  • Add to Cart