ADVH49B827BD AChemBlock

tert-butyl 3-amino-1-oxa-8-azaspiro[3.5]nonane-8-carboxylate

  • Molecular Weight : 242.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-amino-1-oxa-8-azaspiro-3-5-nonane-8-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84650
Availability details for
Quantity
  • Availability
  • Add to Cart