ADVH49B827BF AChemBlock

tert-butyl 3,7-dioxo-2,5-diazaspiro[3.4]octane-5-carboxylate

  • Molecular Weight : 240.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-7-dioxo-2-5-diazaspiro-3-4-octane-5-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84659
Availability details for
Quantity
  • Availability
  • Add to Cart