ADVH49B827C0 AChemBlock

tert-butyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-4,6,7,8-tetrahydropyrazolo[1,5-a][1,4]diazepine-5-carboxylate

  • Molecular Weight : 363.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-4-6-7-8-tetrahydropyrazolo-1-5-a-1-4-diazepine-5-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84667
Availability details for
Quantity
  • Availability
  • Add to Cart