ADVH49B827C1 AChemBlock

tert-butyl 3-(2-iodoethyl)-3-methoxy-azetidine-1-carboxylate

  • Molecular Weight : 341.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-2-iodoethyl-3-methoxy-azetidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84668
Availability details for
Quantity
  • Availability
  • Add to Cart