ADVH49B827C4 AChemBlock

tert-butyl 3-(2-aminoethyl)-3-ethoxy-azetidine-1-carboxylate

  • Molecular Weight : 244.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-2-aminoethyl-3-ethoxy-azetidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84672
Availability details for
Quantity
  • Availability
  • Add to Cart