ADVH49B827C5 AChemBlock

tert-butyl 2-fluoro-4-(hydroxymethyl)-8-azaspiro[4.5]decane-8-carboxylate

  • Molecular Weight : 287.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2-fluoro-4-hydroxymethyl-8-azaspiro-4-5-decane-8-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84674
Availability details for
Quantity
  • Availability
  • Add to Cart