ADVH49B827C8 AChemBlock

tert-butyl 2-{[(4-methylbenzenesulfonyl)oxy]methyl}-4H,5H,6H,7H-[1,3]oxazolo[5,4-c]pyridine-5-carboxylate

  • Molecular Weight : 408.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2-4-methylbenzenesulfonyl-oxy-methyl-4h-5h-6h-7h-1-3-oxazolo-5-4-c-pyridine-5-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84679
Availability details for
Quantity
  • Availability
  • Add to Cart