ADVH49B827D5 AChemBlock

tert-butyl (6S)-3-iodo-6-methyl-4,6,7,8-tetrahydropyrazolo[1,5-a][1,4]diazepine-5-carboxylate

  • Molecular Weight : 377.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-6s-3-iodo-6-methyl-4-6-7-8-tetrahydropyrazolo-1-5-a-1-4-diazepine-5-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84714
Availability details for
Quantity
  • Availability
  • Add to Cart