ADVH49B827D8 AChemBlock

tert-butyl (5S)-5-methyl-4,7-diazaspiro[2.5]octane-7-carboxylate

  • Molecular Weight : 226.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-5s-5-methyl-4-7-diazaspiro-2-5-octane-7-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84720
Availability details for
Quantity
  • Availability
  • Add to Cart