ADVH49B827DD AChemBlock

tert-butyl (5R)-5-methyl-2,7-diazaspiro[3.5]nonane-7-carboxylate

  • Molecular Weight : 240.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-5r-5-methyl-2-7-diazaspiro-3-5-nonane-7-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84729
Availability details for
Quantity
  • Availability
  • Add to Cart