ADVH49B827E3 AChemBlock

tert-butyl (4aR,8aS)-rel-3,3-difluoro-2,4,4a,5,6,7,8,8a-octahydro-1,6-naphthyridine-1-carboxylate

  • Molecular Weight : 276.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4ar-8as-rel-3-3-difluoro-2-4-4a-5-6-7-8-8a-octahydro-1-6-naphthyridine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84744
Availability details for
Quantity
  • Availability
  • Add to Cart