ADVH49B827E5 AChemBlock

tert-butyl (3R)-3-(3-hydroxypropyl)-2,3-dihydro-1H-indole-1-carboxylate

  • Molecular Weight : 277.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3r-3-3-hydroxypropyl-2-3-dihydro-1h-indole-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84761
Availability details for
Quantity
  • Availability
  • Add to Cart