ADVH49B827E8 AChemBlock

tert-butyl (3aR,6aR)-rel-3,3-difluoro-2,3a,4,5,6,6a-hexahydropyrrolo[3,4-b]pyrrole-1-carboxylate

  • Molecular Weight : 248.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3ar-6ar-rel-3-3-difluoro-2-3a-4-5-6-6a-hexahydropyrrolo-3-4-b-pyrrole-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84776
Availability details for
Quantity
  • Availability
  • Add to Cart