ADVH49B827EB AChemBlock

tert-butyl (2R)-2-(hydroxymethyl)-2,3-dihydropyrrolo[3,2-c]pyridine-1-carboxylate

  • Molecular Weight : 250.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2r-2-hydroxymethyl-2-3-dihydropyrrolo-3-2-c-pyridine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84801
Availability details for
Quantity
  • Availability
  • Add to Cart