ADVH49B827F2 AChemBlock

tert-butyl (1R,5R,6R)-6-hydroxy-3,8-diazabicyclo[3.2.1]octane-8-carboxylate

  • Molecular Weight : 228.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-1r-5r-6r-6-hydroxy-3-8-diazabicyclo-3-2-1-octane-8-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84822
Availability details for
Quantity
  • Availability
  • Add to Cart