ADVH49B827F5 AChemBlock

rel-1-benzyl 3-ethyl (3R,4S)-4-ethylpyrrolidine-1,3-dicarboxylate

  • Molecular Weight : 305.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/rel-1-benzyl-3-ethyl-3r-4s-4-ethylpyrrolidine-1-3-dicarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84835
Availability details for
Quantity
  • Availability
  • Add to Cart