ADVH49B827F7 AChemBlock

O5-tert-butyl O3-ethyl 4-methyl-2,4,6,7-tetrahydropyrazolo[4,3-c]pyridine-3,5-dicarboxylate

  • Molecular Weight : 309.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/o5-tert-butyl-o3-ethyl-4-methyl-2-4-6-7-tetrahydropyrazolo-4-3-c-pyridine-3-5-dicarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84849
Availability details for
Quantity
  • Availability
  • Add to Cart