ADVH49B827FA AChemBlock

methyl trans-1-amino-3-ethoxycyclobutane-1-carboxylate hydrochloride

  • Molecular Weight : 209.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-trans-1-amino-3-ethoxycyclobutane-1-carboxylate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84876
Availability details for
Quantity
  • Availability
  • Add to Cart