ADVH49B827FC AChemBlock

methyl cis-1-amino-3-ethoxycyclobutanecarboxylate hydrochloride

  • Molecular Weight : 209.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-cis-1-amino-3-ethoxycyclobutanecarboxylate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84884
Availability details for
Quantity
  • Availability
  • Add to Cart