ADVH49B827FD AChemBlock

methyl 8-hydroxy-2-[(4-methoxyphenyl)methyl]-3,4-dihydro-1H-pyrrolo[1,2-a]pyrazine-7-carboxylate

  • Molecular Weight : 316.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-8-hydroxy-2-4-methoxyphenyl-methyl-3-4-dihydro-1h-pyrrolo-1-2-a-pyrazine-7-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84885
Availability details for
Quantity
  • Availability
  • Add to Cart