ADVH49B82803 AChemBlock

methyl 5-bromo-6-chloro-1H-indole-7-carboxylate

  • Molecular Weight : 288.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-5-bromo-6-chloro-1h-indole-7-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84907
Availability details for
Quantity
  • Availability
  • Add to Cart