ADVH49B82804 AChemBlock

methyl 5-(bromomethyl)norpinane-1-carboxylate

  • Molecular Weight : 247.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-5-bromomethyl-norpinane-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84911
Availability details for
Quantity
  • Availability
  • Add to Cart