ADVH49B82806 AChemBlock

methyl 4H,5H,6H,7H-[1,3]thiazolo[5,4-c]pyridine-2-carboxylate hydrochloride

  • Molecular Weight : 234.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-4h-5h-6h-7h-1-3-thiazolo-5-4-c-pyridine-2-carboxylate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84914
Availability details for
Quantity
  • Availability
  • Add to Cart