ADVH49B82807 AChemBlock

methyl 4-formylbicyclo[2.1.1]hexane-1-carboxylate

  • Molecular Weight : 168.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-formylbicyclo-2-1-1-hexane-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84915
Availability details for
Quantity
  • Availability
  • Add to Cart