ADVH49B8280A AChemBlock

methyl 4-chloro-5H,7H-furo[3,4-d]pyrimidine-2-carboxylate

  • Molecular Weight : 214.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-chloro-5h-7h-furo-3-4-d-pyrimidine-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84918
Availability details for
Quantity
  • Availability
  • Add to Cart