ADVH49B8280E AChemBlock

methyl 4-(tert-butoxycarbonylamino)-3-methyl-cyclohexanecarboxylate

  • Molecular Weight : 271.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-tert-butoxycarbonylamino-3-methyl-cyclohexanecarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84925
Availability details for
Quantity
  • Availability
  • Add to Cart